Assay Design
Gene Transcripts
- Gene Symbol
- Clcn4
- Entrez Gene ID
-
60586
- Gene Name
- chloride channel, voltage-sensitive 4
- Gene Aliases
-
Clcn4-2
- Chromosome Location
-
Chr.X: 25016115 - 25082563 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_022198.2 |
NP_071534.2 |
2 - 3 |
245 |
98 |
GenBank mRNA |
BC092209.1 |
- |
2 - 3 |
245 |
98 |
View Full Product Details