Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100536.1 | NP_001094006.1 | 4 - 5 | 414 | 144 |
XM_006238510.2 | XP_006238572.1 | 4 - 5 | 506 | 144 | |
GenBank mRNA | BQ196337.1 | - | 1 - 2 | 144 | |
CK845348.1 | - | 1 - 2 | 144 | ||
DQ359101.1 | - | 4 - 5 | 343 | 144 |