Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005543.1 | NP_001005543.1 | 15 - 16 | 2420 | 116 |
XM_006238646.2 | XP_006238708.1 | 15 - 16 | 2366 | 116 | |
XM_006238647.2 | XP_006238709.1 | 13 - 14 | 1402 | 116 | |
XM_008763993.1 | XP_008762215.1 | 15 - 16 | 2443 | 116 | |
GenBank mRNA | BC081913.1 | - | 15 - 16 | 2420 | 116 |
FQ212041.1 | - | 14 - 15 | 2360 | 116 |