Assay Design



Gene Transcripts

Gene Symbol
Klhl5
Entrez Gene ID
305351
Gene Name
kelch-like family member 5
Gene Aliases
-
Chromosome Location
Chr.14: 44805213 - 44867078 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047093.1 NP_001040558.1 1 - 2 546 82
GenBank mRNA AY641809.1 - 1 - 2 546 82

View Full Product Details