Assay Design



Gene Transcripts

Gene Symbol
Mfrp
Entrez Gene ID
315597
Gene Name
membrane frizzled-related protein
Gene Aliases
-
Chromosome Location
Chr.8: 48437814 - 48443421 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108137.2 NP_001101607.1 4 - 5 595 58
XM_006242933.2 XP_006242995.1 4 - 5 1040 58

View Full Product Details