Assay Design



Gene Transcripts

Gene Symbol
Commd4
Entrez Gene ID
363068
Gene Name
COMM domain containing 4
Gene Aliases
-
Chromosome Location
Chr.8: 61833340 - 61836856 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108762.1 NP_001102232.1 6 - 7 521 75
XM_006243133.2 XP_006243195.1 7 - 8 647 75
GenBank mRNA FQ213793.1 - 6 - 7 787 75

View Full Product Details