Assay Design



Gene Transcripts

Gene Symbol
Commd4
Entrez Gene ID
363068
Gene Name
COMM domain containing 4
Gene Aliases
-
Chromosome Location
Chr.8: 61833340 - 61836856 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108762.1 NP_001102232.1 3 - 4 144 62
XM_006243133.2 XP_006243195.1 4 - 5 270 62
GenBank mRNA FQ213793.1 - 3 - 4 410 62

View Full Product Details