Assay Design



Gene Transcripts

Gene Symbol
Ddx10
Entrez Gene ID
300710
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 10
Gene Aliases
-
Chromosome Location
Chr.8: 57667472 - 57830504 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106820.1 NP_001100290.1 17 - 18 2585 115

View Full Product Details