Assay Design



Gene Transcripts

Gene Symbol
Zc3h12c
Entrez Gene ID
315658
Gene Name
zinc finger CCCH type containing 12C
Gene Aliases
RGD1565078
Chromosome Location
Chr.8: 56639047 - 56696968 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108146.1 NP_001101616.1 5 - 6 1305 91
XM_006243073.2 XP_006243135.1 5 - 6 1525 91

View Full Product Details