Assay Design



Gene Transcripts

Gene Symbol
Edc3
Entrez Gene ID
315708
Gene Name
enhancer of mRNA decapping 3
Gene Aliases
Lsm16, RGD1306898
Chromosome Location
Chr.8: 62482126 - 62527665 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002729908.4 XP_002729954.2 5 - 6 1160 100

View Full Product Details