Assay Design



Gene Transcripts

Gene Symbol
Zw10
Entrez Gene ID
363059
Gene Name
zw10 kinetochore protein
Gene Aliases
-
Chromosome Location
Chr.8: 53365881 - 53390742 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024801.1 NP_001019972.1 5 - 6 659 127
GenBank mRNA BC097452.1 - 5 - 6 659 127

View Full Product Details