Assay Design



Gene Transcripts

Gene Symbol
Peak1
Entrez Gene ID
315686
Gene Name
pseudopodium-enriched atypical kinase 1
Gene Aliases
RGD1312026
Chromosome Location
Chr.8: 60861742 - 61080046 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108149.1 NP_001101619.1 2 - 3 200 100
XM_006243115.2 XP_006243177.1 1 - 2 70 100

View Full Product Details