Assay Design



Gene Transcripts

Gene Symbol
Zc4h2
Entrez Gene ID
367838
Gene Name
zinc finger, C4H2 domain containing
Gene Aliases
RGD1561708
Chromosome Location
Chr.X: 64887978 - 64908682 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126374.1 NP_001119846.1 2 - 3 347 87
GenBank mRNA BC162002.1 - 2 - 3 347 87

View Full Product Details