Assay Design



Gene Transcripts

Gene Symbol
Smpx
Entrez Gene ID
84416
Gene Name
small muscle protein, X-linked
Gene Aliases
-
Chromosome Location
Chr.X: 40030244 - 40086870 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053395.1 NP_445847.1 4 - 5 461 137
XM_006256945.2 XP_006257007.1 4 - 5 430 137
GenBank mRNA AF364071.1 - 4 - 5 461 137

View Full Product Details