Assay Design



Gene Transcripts

Gene Symbol
Tmem47
Entrez Gene ID
501569
Gene Name
transmembrane protein 47
Gene Aliases
RGD1564799
Chromosome Location
Chr.X: 48779110 - 48805644 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109317.1 NP_001102787.1 2 - 3 597 105

View Full Product Details