Assay Design



Gene Transcripts

Gene Symbol
Slc7a3
Entrez Gene ID
29485
Gene Name
solute carrier family 7 (cationic amino acid transporter, y+ system), member 3
Gene Aliases
Atrc3, Cat3, Slc7a2, cat-3
Chromosome Location
Chr.X: 70972767 - 70978952 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017217.1 NP_058913.1 4 - 5 837 128
XM_006257059.2 XP_006257121.1 5 - 6 1167 128
GenBank mRNA AB000113.1 - 4 - 5 837 128

View Full Product Details