Assay Design



Gene Transcripts

Gene Symbol
Smek3
Entrez Gene ID
302756
Gene Name
SMEK homolog 3, suppressor of mek1 (Dictyostelium)
Gene Aliases
RGD1559817, Smek3p
Chromosome Location
Chr.X: 57866391 - 57872193 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006257000.2 XP_006257062.1 2 - 2 547 139

View Full Product Details