Assay Design



Gene Transcripts

Gene Symbol
Fkbp8
Entrez Gene ID
290652
Gene Name
FK506 binding protein 8
Gene Aliases
FKBP-8
Chromosome Location
Chr.16: 20645956 - 20652890 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037180.1 NP_001032257.1 2 - 2 55 98
GenBank mRNA BC107454.1 - 2 - 2 55 98

View Full Product Details