Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108399.1 | NP_001101869.1 | 5 - 6 | 819 | 59 |
XM_006252909.2 | XP_006252971.1 | 6 - 7 | 810 | 59 | |
XM_008771129.1 | XP_008769351.1 | 5 - 6 | 914 | 59 | |
XM_008771130.1 | XP_008769352.1 | 5 - 6 | 672 | 59 |