Assay Design



Gene Transcripts

Gene Symbol
Mbl1
Entrez Gene ID
24548
Gene Name
mannose-binding lectin (protein A) 1
Gene Aliases
Mbpa, Mlb1
Chromosome Location
Chr.16: 18736154 - 18742193 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012599.2 NP_036731.2 1 - 2 64 89
GenBank mRNA BC088159.1 - 1 - 2 64 89
FQ218582.1 - 1 - 2 92 89

View Full Product Details