Assay Design



Gene Transcripts

Gene Symbol
Babam1
Entrez Gene ID
290631
Gene Name
BRISC and BRCA1 A complex member 1
Gene Aliases
Merit40
Chromosome Location
Chr.16: 19792822 - 19798893 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006964.1 NP_001006965.1 2 - 3 484 78
XM_006252857.2 XP_006252919.1 2 - 3 508 78
GenBank mRNA BC083685.1 - 2 - 3 484 78
FQ214886.1 - 2 - 3 475 78
FQ233705.1 - 2 - 3 495 78

View Full Product Details