Assay Design



Gene Transcripts

Gene Symbol
Babam1
Entrez Gene ID
290631
Gene Name
BRISC and BRCA1 A complex member 1
Gene Aliases
Merit40
Chromosome Location
Chr.16: 19792822 - 19798893 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006964.1 NP_001006965.1 6 - 7 772 81
XM_006252857.2 XP_006252919.1 6 - 7 796 81
GenBank mRNA BC083685.1 - 6 - 7 772 81
FQ214886.1 - 6 - 7 763 81

View Full Product Details