Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012878.2 | NP_037010.1 | 2 - 3 | 333 | 66 |
XM_008771065.1 | XP_008769287.1 | 2 - 3 | 336 | 66 | |
GenBank mRNA | BC070507.1 | - | 3 - 4 | 342 | 66 |
M81231.1 | - | 3 - 4 | 340 | 66 |