Assay Design



Gene Transcripts

Gene Symbol
Sec61a1
Entrez Gene ID
80843
Gene Name
Sec61 alpha 1 subunit (S. cerevisiae)
Gene Aliases
Sec61a
Chromosome Location
Chr.4: 120453577 - 120467932 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199256.2 NP_954865.1 6 - 7 568 76
GenBank mRNA BC094530.1 - 6 - 7 568 76
M96630.1 - 6 - 7 559 76

View Full Product Details