Assay Design



Gene Transcripts

Gene Symbol
Elmod1
Entrez Gene ID
315670
Gene Name
ELMO/CED-12 domain containing 1
Gene Aliases
RGD1564186
Chromosome Location
Chr.8: 58486078 - 58542888 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191579.1 NP_001178508.1 3 - 4 355 97
XM_006243083.2 XP_006243145.1 3 - 4 399 97
XM_008766269.1 XP_008764491.1 3 - 4 437 97

View Full Product Details