Assay Design
Gene Transcripts
- Gene Symbol
- Idh3a
- Entrez Gene ID
-
114096
- Gene Name
- isocitrate dehydrogenase 3 (NAD+) alpha
- Gene Aliases
-
-
- Chromosome Location
-
Chr.8: 59164601 - 59183899 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_053638.1 |
NP_446090.1 |
10 - 11 |
1071 |
62 |
GenBank mRNA |
AB047541.1 |
- |
10 - 11 |
1071 |
62 |
View Full Product Details