Assay Design
Gene Transcripts
- Gene Symbol
- Coro2b
- Entrez Gene ID
-
300768
- Gene Name
- coronin, actin binding protein, 2B
- Gene Aliases
-
-
- Chromosome Location
-
Chr.8: 67374617 - 67492097 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_217183.8 |
XP_217183.6 |
6 - 7 |
856 |
84 |
View Full Product Details