Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013168.2 | NP_037300.2 | 3 - 4 | 283 | 138 |
XM_006242896.1 | XP_006242958.1 | 3 - 4 | 369 | 138 | |
XM_006242897.1 | XP_006242959.1 | 3 - 4 | 369 | 138 | |
GenBank mRNA | BC088162.1 | - | 3 - 4 | 283 | 138 |
FQ226062.1 | - | 3 - 4 | 206 | 138 | |
FQ231091.1 | - | 3 - 4 | 267 | 138 | |
X06827.1 | - | 3 - 4 | 158 | 138 | |
Y12006.1 | - | 3 - 4 | 153 | 138 |