Assay Design



Gene Transcripts

Gene Symbol
Fxyd2
Entrez Gene ID
29639
Gene Name
FXYD domain-containing ion transport regulator 2
Gene Aliases
ATP1C, Atp1g1, GNAKATP
Chromosome Location
Chr.8: 49710328 - 49717492 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006242912.2 XP_006242974.1 1 - 2 165 114

View Full Product Details