Assay Design



Gene Transcripts

Gene Symbol
Pou2f3
Entrez Gene ID
116544
Gene Name
POU class 2 homeobox 3
Gene Aliases
-
Chromosome Location
Chr.8: 47412063 - 47495042 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105745.1 NP_001099215.1 3 - 4 200 75
GenBank mRNA L23862.1 - 3 - 4 170 75

View Full Product Details