Assay Design



Gene Transcripts

Gene Symbol
Serpinb11
Entrez Gene ID
304689
Gene Name
serpin peptidase inhibitor, clade B (ovalbumin), member 11
Gene Aliases
-
Chromosome Location
Chr.13: 27217385 - 27257181 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107167.1 NP_001100637.1 4 - 5 482 99
XM_006249632.2 XP_006249694.2 3 - 4 632 99
XM_006249633.2 XP_006249695.2 3 - 4 334 99

View Full Product Details