Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107167.1 | NP_001100637.1 | 7 - 8 | 860 | 141 |
XM_006249632.2 | XP_006249694.2 | 6 - 7 | 1010 | 141 | |
XM_006249633.2 | XP_006249695.2 | 6 - 7 | 712 | 141 | |
XM_008769453.1 | XP_008767675.1 | 3 - 4 | 503 | 141 |