Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008312.1 | NP_001008313.1 | 7 - 8 | 969 | 103 |
GenBank mRNA | BC085853.1 | - | 7 - 8 | 969 | 103 |
EU220431.1 | - | 7 - 8 | 915 | 103 | |
FQ222836.1 | - | 1 - 2 | 52 | 103 |