Assay Design



Gene Transcripts

Gene Symbol
Gdap1l1
Entrez Gene ID
311616
Gene Name
ganglioside-induced differentiation-associated protein 1-like 1
Gene Aliases
Gdap1l
Chromosome Location
Chr.3: 159823755 - 159843238 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107798.2 NP_001101268.1 3 - 4 562 90
GenBank mRNA BC168159.1 - 3 - 4 562 90
FQ212886.1 - 3 - 4 619 90

View Full Product Details