Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145670.4 | NP_663703.3 | 4 - 5 | 884 | 93 |
XM_008762466.1 | XP_008760688.1 | 4 - 5 | 884 | 93 | |
GenBank mRNA | AB029897.1 | - | 2 - 3 | 392 | 93 |
AB180269.1 | - | 2 - 3 | 392 | 93 | |
AB180270.1 | - | 2 - 3 | 392 | 93 | |
AB180271.1 | - | 2 - 3 | 392 | 93 | |
AB180272.1 | - | 2 - 3 | 392 | 93 | |
BC094306.1 | - | 2 - 3 | 380 | 93 | |
BC103480.1 | - | 3 - 4 | 782 | 93 |