Assay Design



Gene Transcripts

Gene Symbol
Fam83d
Entrez Gene ID
311598
Gene Name
family with sequence similarity 83, member D
Gene Aliases
RGD1565583
Chromosome Location
Chr.3: 155269344 - 155288612 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107796.1 NP_001101266.1 3 - 4 785 101

View Full Product Details