Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025768.1 | NP_001020939.1 | 8 - 9 | 788 | 83 |
XM_006235375.2 | XP_006235437.1 | 8 - 9 | 1098 | 83 | |
XM_006235376.2 | XP_006235438.1 | 8 - 9 | 1769 | 83 | |
XM_006235377.2 | XP_006235439.1 | 8 - 9 | 1861 | 83 | |
XM_006235378.2 | XP_006235440.1 | 8 - 9 | 1098 | 83 | |
GenBank mRNA | BC098027.1 | - | 8 - 9 | 788 | 83 |