Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024381.2 | NP_077357.2 | 9 - 10 | 858 | 158 |
XM_006256987.2 | XP_006257049.1 | 10 - 11 | 910 | 176 | |
XM_006256988.2 | XP_006257050.1 | 9 - 10 | 892 | 158 | |
XM_008773274.1 | XP_008771496.1 | 11 - 12 | 975 | 176 | |
XM_008773275.1 | XP_008771497.1 | 10 - 11 | 957 | 158 | |
XM_008773276.1 | XP_008771498.1 | 11 - 12 | 975 | 176 | |
XM_008773278.1 | XP_008771500.1 | 11 - 12 | 974 | 176 | |
GenBank mRNA | D16102.1 | - | 9 - 10 | 861 | 158 |