Assay Design



Gene Transcripts

Gene Symbol
Tbl1x
Entrez Gene ID
302711
Gene Name
transducin (beta)-like 1 X-linked
Gene Aliases
RGD1563868
Chromosome Location
Chr.X: 44959883 - 44988551 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106964.1 NP_001100434.1 - 1315 69
GenBank mRNA BC167084.1 - - 1410 69

View Full Product Details