Assay Design



Gene Transcripts

Gene Symbol
Rogdi
Entrez Gene ID
287061
Gene Name
rogdi homolog (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.10: 10761477 - 10766096 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024864.1 NP_001020035.1 6 - 7 619 74
GenBank mRNA BC083789.1 - 4 - 5 1050 74
BC098001.1 - 6 - 7 619 74

View Full Product Details