Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106973.1 | NP_001100443.1 | 7 - 8 | 690 | 116 |
XM_006245755.1 | XP_006245817.1 | 8 - 9 | 1029 | 116 | |
GenBank mRNA | BC168922.1 | - | 7 - 8 | 646 | 116 |
FQ232490.1 | - | 2 - 3 | 184 | 116 |