Assay Design
Gene Transcripts
- Gene Symbol
- Grin2a
- Entrez Gene ID
-
24409
- Gene Name
- glutamate receptor, ionotropic, N-methyl D-aspartate 2A
- Gene Aliases
-
GluN2A, NMDAR2A, NR2A
- Chromosome Location
-
Chr.10: 5707806 - 6119897 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_012573.3 |
NP_036705.3 |
6 - 7 |
1712 |
84 |
GenBank mRNA |
D13211.1 |
- |
5 - 6 |
1505 |
84 |
M91561.1 |
- |
6 - 7 |
1712 |
84 |
View Full Product Details