Assay Design
Gene Transcripts
- Gene Symbol
- Atp2b2
- Entrez Gene ID
-
24215
- Gene Name
- ATPase, Ca++ transporting, plasma membrane 2
- Gene Aliases
-
PMCA2
- Chromosome Location
-
Chr.4: 145704779 - 145948997 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_012508.5 |
NP_036640.1 |
- |
744 |
96 |
GenBank mRNA |
J03754.1 |
- |
- |
750 |
96 |
View Full Product Details