Assay Design



Gene Transcripts

Gene Symbol
Ccdc37
Entrez Gene ID
297444
Gene Name
coiled-coil domain containing 37
Gene Aliases
RGD1305768
Chromosome Location
Chr.4: 122314475 - 122339007 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006236885.2 XP_006236947.1 1 - 2 784 66

View Full Product Details