Assay Design



Gene Transcripts

Gene Symbol
Slc41a3
Entrez Gene ID
641603
Gene Name
solute carrier family 41, member 3
Gene Aliases
-
Chromosome Location
Chr.4: 123467552 - 123513451 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037492.1 NP_001032569.1 6 - 7 1025 102
XM_006236871.2 XP_006236933.1 7 - 8 1035 102
XM_006236872.2 XP_006236934.1 7 - 8 1092 102
XM_006236873.1 XP_006236935.1 7 - 8 1409 102
XM_006236874.2 XP_006236936.1 6 - 7 964 102
XM_006236875.2 XP_006236937.1 7 - 8 1241 102
XM_006236876.2 XP_006236938.1 6 - 7 636 102
XM_006236877.2 XP_006236939.1 6 - 7 1381 102
XM_006236878.2 XP_006236940.1 6 - 7 682 102
XM_006236879.2 XP_006236941.1 6 - 7 677 102
XM_008763134.1 XP_008761356.1 7 - 8 1236 102
GenBank mRNA BC104698.1 - 6 - 7 1025 102

View Full Product Details