Assay Design



Gene Transcripts

Gene Symbol
Slc41a3
Entrez Gene ID
641603
Gene Name
solute carrier family 41, member 3
Gene Aliases
-
Chromosome Location
Chr.4: 123467552 - 123513451 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037492.1 NP_001032569.1 8 - 9 1246 114
XM_006236871.2 XP_006236933.1 9 - 10 1256 114
XM_006236872.2 XP_006236934.1 9 - 10 1313 114
XM_006236873.1 XP_006236935.1 9 - 10 1630 114
XM_006236874.2 XP_006236936.1 8 - 9 1185 114
XM_006236875.2 XP_006236937.1 9 - 10 1462 114
XM_006236876.2 XP_006236938.1 8 - 9 857 114
XM_006236877.2 XP_006236939.1 8 - 9 1602 114
XM_006236878.2 XP_006236940.1 8 - 9 903 114
XM_006236879.2 XP_006236941.1 8 - 9 898 114
XM_008763134.1 XP_008761356.1 9 - 10 1457 114
GenBank mRNA BC104698.1 - 8 - 9 1246 114

View Full Product Details