Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010956.2 | NP_001010956.1 | 6 - 7 | 966 | 137 |
NM_001033931.1 | NP_001029103.1 | 6 - 7 | 966 | 137 | |
GenBank mRNA | AY862307.1 | - | 6 - 7 | 793 | 137 |
AY862308.1 | - | 6 - 7 | 761 | 137 |