Assay Design



Gene Transcripts

Gene Symbol
Vps4b
Entrez Gene ID
360834
Gene Name
vacuolar protein sorting 4 homolog B (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.13: 26825261 - 26850460 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025716.1 NP_001020887.1 4 - 5 554 87
GenBank mRNA BC099128.1 - 4 - 5 554 87

View Full Product Details