Assay Design



Gene Transcripts

Gene Symbol
Capn9
Entrez Gene ID
116694
Gene Name
calpain 9
Gene Aliases
Ncl-4
Chromosome Location
Chr.19: 57341938 - 57378901 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271140.1 NP_001258069.1 5 - 6 733 68
GenBank mRNA U89514.1 - 5 - 6 648 68

View Full Product Details