Assay Design



Gene Transcripts

Gene Symbol
Chst4
Entrez Gene ID
307838
Gene Name
carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 4
Gene Aliases
-
Chromosome Location
Chr.19: 41630282 - 41642373 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107427.1 NP_001100897.1 1 - 2 98 65

View Full Product Details